missing translation for 'onlineSavingsMsg'
Learn More

Minaprine dihydrochloride, MedChemExpress

Product Code. 16408486
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408486 Supplier Medchem Express Supplier No. HYB0884A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Minaprine dihydrochloride is a reversible inhibitor of MAO-A; weakly inhibit acetylcholinesterase; an antidepressant for treatment of depression.

Minaprine dihydrochloride is a reversible inhibitor of MAO-A

Specifications

Chemical Name or Material Minaprine dihydrochloride
Physical Form Solid
CAS 25953-17-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H24Cl2N4O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 100 mg/mL (269.32 mM) ∣H2O : 100 mg/mL (269.32 mM; Need ultrasonic)
SMILES [H]Cl.CC1=CC(C2=CC=CC=C2)=NN=C1NCCN3CCOCC3.[H]Cl
Molecular Weight (g/mol) 371.3
Formula Weight 371.3
Percent Purity 98.82%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.