missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Mineralocorticoid R/NR3C2 Antibody (PCRP-NR3C2-1E3), Novus Biologicals™
Mouse Monoclonal Antibody
232.00 € - 462.00 €
Specifications
| Antigen | Mineralocorticoid R/NR3C2 |
|---|---|
| Clone | PCRP-NR3C2-1E3 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043666
|
Novus Biologicals™
NBP3-26819-100ug |
100 μg |
462.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043953
|
Novus Biologicals™
NBP3-26819-20ug |
20 μg |
232.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mineralocorticoid R/NR3C2 Monoclonal antibody specifically detects Mineralocorticoid R/NR3C2 in Human samples. It is validated for Flow CytometrySpecifications
| Mineralocorticoid R/NR3C2 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 4306 | |
| IgG1 | |
| Protein A or G purified |
| PCRP-NR3C2-1E3 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
| Recombinant fragment (around aa601-673) of human Mineralocorticoid R/NR3C2 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title