missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Minerals Modified Glutamate

Product Code. 12736989
Click to view available options
Media Type:
MMGA (Bottled)
MMGA 2X Strength + DurhamTube
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 12736989

Brand: Thermo Scientific™ BO0542E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enrich and identify members of the coliform group of bacteria in food and water samples with these ready-prepared bottles.

Enrich and identify members of the coliform group of bacteria in food and water samples with Thermo Scientific™ Oxoid™ Minerals Modified Glutamate ready-prepared bottled medium.The mineral content of the medium allows for the superior cultivation and enumeration of coliforms.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Minerals Modified Glutamate Medium.

  • Convenient: pre-filled bottles are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.
  • Superior performance over MacConkey Broth in the detection of E. coli2.
  • The medium of choice for detection of small numbers of E. coli in chlorinated water.
  • Effectively isolates E. coli from sea water.

Minerals Modified Glutamate Medium was developed by Oxoid Laboratories and Metropolitan Water Board Laboratories, and is based upon the formulation described by Gray1

TRUSTED_SUSTAINABILITY

Specifications

Color Purple, Clear
Content And Storage 2 to 25°C
Format Universal Bottle
Manufacturing Origin United Kingdom
Media Type MMGA 2X Strength + DurhamTube
Product Type Agar
Fill Weight 10.0 + 0.3g
Packaging Type 24 universal bottles with screw cap closures in a box
Quantity 24/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.