missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Minimum Essential Medium Eagle (modified) 1X- without L-Glutamine

Product Code. 17358573 Shop All MP Biomedicals Products
Change view
Click to view available options
Form:
Liquid
Unit Size:
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form Unit Size
17358573 Liquid 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17358573 Supplier MP Biomedicals™ Supplier No. 091210254X6
  • ONLINE EXCLUSIVE PRICE
    141.00 €
    133.35 € / 3 L
    Save 7.65 €
    5% Off
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MEM cell culture media that can be used with either Earle's or Hanks' salts.

MEM, originally prepared by Harry Eagle, is one of the most popular cell culture media. Upon his attempts to cultivate normal mammalian fibroblasts and certain HeLa cell subtypes, it was revealed that the nutritional needs of these cell types could not be met by BME. Further studies led to the development of MEM, incorporating specific modifications such as higher amino acid concentrations for the cultivation of fastidious cells. MP Biomedical™ offers MEM with either Earle's or Hanks' salts.

MP Biomedical™ MEM may be used to support the growth of:

  • Cells in monolayers
  • Cells in suspension
  • Wide variety of other cell types with proper supplementation

Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Specifications

Form Liquid
Product Type Cell Culture Medium
With Additives Phenol Red
Without Additives No Glucose, No Glutamine, No Sodium Pyruvate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.