missing translation for 'onlineSavingsMsg'
Learn More

Minocycline hydrochloride

Product Code. 15427789
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427789 500 mg 500mg
15417789 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427789 Supplier Thermo Scientific Alfa Aesar Supplier No. J66429.ME
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13614-98-7 | C23H28ClN3O7 | 493.94 g/mol

A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13614-98-7
Molecular Formula C23H28ClN3O7
Molecular Weight (g/mol) 493.94
MDL Number MFCD00083669
InChI Key KDLQIOPKJDNQIM-WUURTAMISA-N
PubChem CID 54685925
IUPAC Name hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
SMILES [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

Type Minocycline Hydrochloride
Physical Form Powder
Melting Point 205°C to 210°C
Color Yellow
Quantity 500 mg
Flash Point >110°C (230°F)
Boiling Point 813°C
Merck Index 14,6202
Solubility Information Freely soluble in water
Formula Weight 493.94
Chemical Name or Material Minocycline hydrochloride, 890 to 950μg/mg
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.