missing translation for 'onlineSavingsMsg'
Learn More

Minocycline hydrochloride, MedChemExpress

Product Code. 16433347
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16433347 50 mg 50mg
16427296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16433347 Supplier Medchem Express Supplier No. HY17412
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Minocycline hydrochloride is an orally active, potent and BBB-penetrated semi-synthetic tetracycline antibiotic.

Minocycline hydrochloride is a hypoxia-inducible factor (HIF)-1α inhibitor. Minocycline hydrochloride shows anti-cancer, anti-inflammatory, and glutamate antagonist effects. Minocycline hydrochloride reduces glutamate neurotransmission and shows neuroprotective properties and antidepressant effects. Minocycline hydrochloride inhibits bacterial protein synthesis through binding with the 30S subunit of the bacterial ribosome, resulting in a bacteriostatic effect.

Specifications

Chemical Name or Material Minocycline hydrochloride
CAS 13614-98-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H28ClN3O7
Quantity 50 mg
Solubility Information H2O : 10 mg/mL (20.25 mM; Need ultrasonic) ∣DMSO : 0.49 mg/mL (0.99 mM; Need ultrasonic and warming)
SMILES OC1=C(C(N)=O)C([C@@]2(O)C(O)=C3C(C4=C(O)C=CC(N(C)C)=C4C[C@@]3([H])C[C@@]2([H])[C@@H]1N(C)C)=O)=O.Cl
Molecular Weight (g/mol) 493.94
Formula Weight 493.94
Percent Purity 98.47%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.