missing translation for 'onlineSavingsMsg'
Learn More

Minoxidil, MedChemExpress

Product Code. 16482049
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482049 1 g 1g
16440778 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482049 Supplier Medchem Express Supplier No. HYB0112
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Minoxidil (U10858) is an ATP-sensitive potassium (KATP) channel opener, a potent oral antihypertensive agent and a peripheral vasodilator that promotes vasodilation also affects hair growth. Minoxidil is also a potent inhibitor of soybean lipoxygenaseare with an IC50 of 20 μM.

Minoxidil (U10858) is an ATP-sensitive potassium (KATP) channel opener, a potent oral antihypertensive agent and a peripheral vasodilator that promotes vasodilation also affects hair growth. Minoxidil is also a potent inhibitor of soybean lipoxygenaseare with an IC50 of 20 μM.

Specifications

Chemical Name or Material Minoxidil
Physical Form Powder
CAS 38304-91-5
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C9H15N5O
For Use With (Application) COVID-19-immunoregulation
Synonym U10858
Solubility Information Ethanol : 7.14 mg/mL (34.12 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.78 mM; ultrasonic and warming and heat to 50°C)
SMILES NC1=NC(N2CCCCC2)=CC(N)=[N+]1[O-]
Molecular Weight (g/mol) 209.25
Formula Weight 209.25
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.