missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
The MiRP3 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Tekniske data
Tekniske data
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC24246 and based on accession number NM_8671. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens potassium voltage-gated channel, Isk-related family, member 4 (KCNE4), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?