Learn More
MIS18BP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552306
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QHQRILLPSF QDSEDDDDIL PNMDKNPTTP SSVIFPLVKT PQCQHVSPGM LGSINRNDCD KYVFRMQKYH KSNGGIVWGN IKKKLVETDF STPTPRRKTP FNTDLGENSG IGKLFTNAVE SLDE Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 61%.
MIS18BP1 is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.Specifications
MIS18BP1 | |
Polyclonal | |
Unconjugated | |
MIS18BP1 | |
6720403H11; C14orf106; C79407; HSA242977; HsKNL-2; KIAA1903; kinetochore null 2 homolog; kinetochore-associated protein KNL-2 homolog; Knl2; M18BP1; MIS18 binding protein 1; mis18-binding protein 1; Mis18bp1; mKIAA1903; P243; putative protein p243 which interacts with transcription factor Sp1; RGD1305497 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55320 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P0N0 | |
MIS18BP1 | |
Recombinant protein corresponding to Human MIS18BP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only