missing translation for 'onlineSavingsMsg'
Learn More

MISA Standard 5: Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb @ 100 μg/mL in 2% HNO3, VHG™

Product Code. 30003103
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003103 100 mL 100mL
30003158 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30003103 Leverandør VHG Labs Leverandørnr. VHGMISA5100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

MISA Standard 5: Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb @ 100 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Holmium/Erbium/Samarium/Thulium/Caesium/Terbium/Scandium/Dysprosium/Europium/Neodymium/Ytterbium/Lanthanum/Praseodymium/Thorium/Yttrium/Gadolinium/Lutetium
CAS 7440-60-0, 7440-52-0, 7440-19-9, 7440-30-4, 7440-46-2, 7440-27-9
Methods ICP/ICP-MS
Analyte or Component Names Holmium/Erbium/Samarium/Thulium/Caesium/Terbium/Scandium/Dysprosium/Europium/Neodymium/Ytterbium/Lanthanum/Praseodymium/Thorium/Yttrium/Gadolinium/Lutetium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Holmium 99.88 μg/mL/Erbium 99.94 μg/mL/Samarium 99.95 μg/mL/Thulium 99.96 μg/mL/Caesium 99.97 μg/mL/Terbium 99.97 μg/mL/Scandium 99.98 μg/mL/Dysprosium 99.99 μg/mL/Europium 100 μg/mL/Neodymium 100 μg/mL/Ytterbium 100 μg/mL/Lanthanum 100.1 μg/mL/Praseodymium 100.1 μg/mL/Thorium 100.1 μg/mL/Yttrium 100.1 μg/mL/Gadolinium 100.2 μg/mL/Lutetium 100.2 μg/mL
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.