missing translation for 'onlineSavingsMsg'
Learn More

Mitiglinide calcium, MedChemExpress

Product Code. 16471846
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16471846

Brand: Medchem Express HY17398

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mitiglinide Calcium (KAD-1229 anhydrous), an insulinotropic agent, is an ATP-sensitive K+ (KATP) channel antagonist. Mitiglinide Calcium is highly specific to the Kir6.2/SUR1 complex (the pancreatic beta-cell KATP channel). Mitiglinide Calcium can be used for the research of type 2 diabetes.

Mitiglinide Calcium (KAD-1229 anhydrous), an insulinotropic agent, is an ATP-sensitive K+ (KATP) channel antagonist. Mitiglinide Calcium is highly specific to the Kir6.2/SUR1 complex (the pancreatic beta-cell KATP channel). Mitiglinide Calcium can be used for the research of type 2 diabetes.

Specifications

Chemical Name or Material Mitiglinide calcium
Physical Form Solid
CAS 145525-41-3
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C19H24NO3.1/2Ca
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym KAD-1229 anhydrousS21403 anhydrous
Solubility Information DMSO : 5 mg/mL (14.95 mM; Need ultrasonic)
SMILES O=C([O-])[C@H](CC(N1C[C@](CCCC2)([H])[C@]2([H])C1)=O)CC3=CC=CC=C3.[0.5Ca2+]
Molecular Weight (g/mol) 334.44
Formula Weight 334.44
Percent Purity 98.7%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.