missing translation for 'onlineSavingsMsg'
Learn More

Mitomycin C, Thermo Scientific Chemicals

Product Code. 15415459
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415459 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415459 Supplier Thermo Scientific Alfa Aesar Supplier No. J63193.#0
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mitomycin C, CAS # 50-07-7, is a methylazirinopyrroloindoledione compound showing antineoplastic and antibiotic activities. It is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria.

Mitomycin C is used as a chemotherapeutic agent due to its antitumor activity. It is used to treat upper gastro-intestinal cancers. It serves as an antineoplastic agent which inhibits DNA synthesis and induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Mitomycin C is a methylazirinopyrroloindoledione compound isolated from the bacterium Streptomyces caespitosus, and other Streptomyces bacterial species that has antineoplastic and antibiotic activities
• This compound acts as an inhibitor of DNA synthesis, nuclear division, as well as an apoptosis inducer. It can create oxygen radicals, and produce inter-strand DNA crosslinks, inhibiting DNA synthesis
• Mitomycin C shows alkylating activity and mainly targets the guanine nucleoside sequence 5′-CpG-3′. It inhibits DNA synthesis by covalently reacting with DNA, forming crosslinks between complementary strands of DNA. Consequently, this interaction prevents the separation of complementary DNA strands, inhibiting DNA replication

Application

• Mitomycin C is active against a broad-spectrum of gram-positive, gram-negative, and acid-fast bacilli bacteria
• Mitomycin C has strong antineoplastic activity, mainly against Ehrlich ascites tumor cell lines
• This compound can inhibit B and T cells and macrophage proliferation. It can also impair antigen presentation, as well as the secretion of interferon-γ, TNF- α, and IL-2
• This compound is used to generate mitotically inactive feeder cells in cell culture systems, such as the mitotically inactive fibroblasts used in embryonic stem cell systems
• It has been used to treat basal-like cancer cell line in vitro experiments
• It is particularly toxic to hypoxic cells inhibiting RNA and protein synthesis at high concentrations

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.332
MDL Number MFCD00078109
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Type Mitomycin C
Physical Form Powder
Melting Point 360°C
Color Blue to Purple
Quantity 5 mg
UN Number UN2811
Beilstein 7231816
Merck Index 14,6215
Solubility Information Soluble in methanol at 10mg/ml; Also soluble in water at 1mg/ml with warming; Soluble in DMSO at ≈15mg/ml
Formula Weight 334.33
Chemical Name or Material Mitomycin C
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.