missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ MitoProbe™ DiIC1(5) Assay Kit - For Flow Cytometry - 100 Assays
Click to view available options
Quantity:
100 kit
Unit Size:
Each
Description
The MitoProbe™ DiIC1(5) Assay Kit provides solutions of the membrane-potential-sensitive cyanine dye DiIC1(5), and a mitochondrial membrane-potential disrupter, CCCP, for the study of mitochondrial membrane potential. DiIC1(5) penetrates the cytosol of eukaryotic cells and accumulates primarily in mitochondria with active membrane potentials, producing bright, far-red fluorescence. DiIC1(5) staining intensity decreases with decreased mitochondrial membrane potential.
View a selection guide for all apoptosis assays for flow cytometry.
View a selection guide for all apoptosis assays for flow cytometry.
Specifications
Specifications
| Content And Storage | Contains 1 vial of DiIC1(5) ( 625 μL, 10 μM in DMSO), and 1 vial of CCCP: (125 μL, 50 mM DMSO). Store at 2–6°C, protected from light. Do not freeze. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Flow Cytometry Assay Kit |
| Dye Type | DiIC1(5) |
| Emission | 633/635 |
| Format | Tube(s) |
| Product Line | MitoProbe |
| Quantity | 100 kit |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction