Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems
Product Code. 30040273 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
Port Detail:
4 x Top-mounted Ports and Tubing
Powder Port
Unit Size:
Pack of 15
Pack of 3
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Port Detail unitSize
30040273 50 L Powder Port Pack of 3
30040271 50 L 4 x Top-mounted Ports and Tubing Pack of 15
30040272 100 L 4 x Top-mounted Ports and Tubing Pack of 15
30040270 200 L 4 x Top-mounted Ports and Tubing Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30040273 Supplier Thermo Scientific™ Supplier No. SH30687.01
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products.
Recommended for:
  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 L
Port Detail Powder Port
Tubing Style TPE