missing translation for 'onlineSavingsMsg'
Learn More

MK-0249, MedChemExpress

Product Code. 16483129
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483129 1 mg 1mg
16412348 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16483129 Supplier Medchem Express Supplier No. HYU00076
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress MK-0249 (Compound 1) is a potent, selective and orally active histamine H3 receptor antagonist, with an IC50 of 1.7 nM for human H3.

MK-0249 (Compound 1) is a potent, selective and orally active histamine H3 receptor antagonist, with an IC50 of 1.7 nM for human H3.

Specifications

Chemical Name or Material MK-0249
CAS 862309-06-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H24F3N3O2
Quantity 1 mg
Solubility Information DMSO : 50 mg/mL (115.89 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=C(OCCCN3CCCC3)C=C2)C(C)=NC4=C1C(C(F)(F)F)=CC=C4
Molecular Weight (g/mol) 431.45
Formula Weight 431.45
Percent Purity 96.94%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.