missing translation for 'onlineSavingsMsg'
Learn More

MK 0893, MedChemExpress

Product Code. 16493987
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493987 50 mg 50mg
16460538 5 mg 5mg
16408078 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493987 Supplier Medchem Express Supplier No. HY50663
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress MK 0893 is a potent and selective glucagon receptor antagonist with an IC50 of 6.6 nM.

MK 0893 is a potent and selective glucagon receptor antagonist with an IC50 of 6.6 nM.

Specifications

Chemical Name or Material MK 0893
CAS 870823-12-4
Color White
Purity Grade Notes Research
Molecular Formula C32H27Cl2N3O4
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (56.64 mM; Need ultrasonic)
SMILES ClC1=CC(C2=NN(C(C3=CC4=C(C=C3)C=C(OC)C=C4)=C2)[C@H](C5=CC=C(C=C5)C(NCCC(O)=O)=O)C)=CC(Cl)=C1
Molecular Weight (g/mol) 588.48
Formula Weight 588.48
Percent Purity 98.27%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.