missing translation for 'onlineSavingsMsg'
Learn More

MK-2206 dihydrochloride, MedChemExpress

Product Code. 16496587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496587 5 mg 5mg
16460976 200 mg 200mg
16450237 50 mg 50mg
16423868 10 mg 10mg
16454896 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496587 Supplier Medchem Express Supplier No. HY10358
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy.

MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy.

Specifications

Chemical Name or Material MK-2206 dihydrochloride
CAS 1032350-13-2
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C25H23Cl2N5O
Quantity 5 mg
Synonym MK-2206 (2HCl)
Solubility Information DMSO : 16.67 mg/mL (34.70 mM; ultrasonic and warming and heat to 70°C) ∣H2O : 3.57 mg/mL (7.43 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1N2C(C3=CC(C4=CC=CC=C4)=C(N=C3C=C2)C5=CC=C(C6(N)CCC6)C=C5)=NN1.Cl.Cl
Molecular Weight (g/mol) 480.39
Formula Weight 480.39
Percent Purity 98.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.