missing translation for 'onlineSavingsMsg'
Learn More

MK-2461, MedChemExpress

Product Code. 16400356
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400356

Brand: Medchem Express HY50703

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress MK-2461 is a novel ATP-competitive multitargeted inhibitor of activated c-Met with a mean IC50 of 2.5 nM.

MK-2461 is a novel ATP-competitive multitargeted inhibitor of activated c-Met with a mean IC50 of 2.5 nM.

Specifications

Chemical Name or Material MK-2461
Physical Form Solid
CAS 917879-39-1
Color Yellow
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C24H25N5O5S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 31 mg/mL (62.56 mM)
SMILES O=S(N(C)C[C@H]1OCCOC1)(NC2=CC=C3C=CC(C(C(C3=C2)=O)=C4)=NC=C4C5=CN(N=C5)C)=O
Molecular Weight (g/mol) 495.55
Formula Weight 495.55
Percent Purity 99.01%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.