missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MK-2894 sodium salt, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
MK-2894 sodium salt is a potent, selective, orally active and high affinity (Ki=0.56 nM) full antagonist against E prostanoid receptor 4 (EP4 receptor) (IC50=2.5 nM). MK-2894 sodium salt possesses potent anti-inflammatory activity in animal models of pain/inflammation and can be used for the research of arthritis.
Specifications
Specifications
| Chemical Name or Material | MK-2894 sodium salt |
| Physical Form | Solid |
| CAS | 1006036-88-9 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C25H21F3NNaO3S |
| For Use With (Application) | COVID-19-immunoregulation |
| SMILES | O=C(O[Na])C1=CC=C(C2(NC(C3=C(C)SC(C)=C3CC4=CC=C(C(F)(F)F)C=C4)=O)CC2)C=C1 |
| Molecular Weight (g/mol) | 495.49 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction