missing translation for 'onlineSavingsMsg'
Learn More

(+)-MK 801 Hydrogen Maleate, TRC

Product Code. 18002520
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18002520 10 mg 10mg
18032220 25 mg 25mg
18092510 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18002520 Supplier Toronto Research Chemicals Supplier No. TRCM42495010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77086-22-7
Molecular Formula C16 H15 N . C4 H4 O4
Molecular Weight (g/mol) 337.37
Synonym (5S)-10,11-Dihydro-5-methyl-5H-dibenzo[a,d]cyclohepten-5,10-imine, (2Z)-2-butenedioate (1:1), (5S)-10,11-Dihydro-5-methyl-5H-dibenzo[a,d]cyclohepten-5,10-imine (Z)-2-Butenedioate (1:1), Dizocilpine Hydrogen Maleate, Dizocilpine Maleate
SMILES C[C@]12N[C@H](Cc3ccccc13)c4ccccc24.OC(=O)\C=C/C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H15N.C4H4O4/c1-16-13-8-4-2-6-11(13)10-15(17-16)12-7-3-5-9-14(12)16;5-3(6)1-2-4(7)8/h2-9,15,17H,10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t15-,16+;/m1./s1
Formula Weight 337.1314
Chemical Name or Material (+)-MK 801 Hydrogen Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.