missing translation for 'onlineSavingsMsg'
Learn More

ML-18, MedChemExpress

Product Code. 16499996
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499996 50 mg 50mg
16474696 100 mg 100mg
16453608 10 mg 10mg
16452786 25 mg 25mg
16426337 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499996 Supplier Medchem Express Supplier No. HY101844
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.

ML-18 is a non-peptide bombesin receptor subtype-3 (BRS-3) antagonist with an IC50 of 4.8 μM.

Specifications

Chemical Name or Material ML-18
CAS 1422269-30-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C32H35N5O5
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (175.55 mM)
SMILES O=C([C@@H](NC(NC1=CC=C([N+]([O-])=O)C=C1)=O)CC2=CNC3=CC=CC=C23)NCC4(C5=CC=C(OC)C=C5)CCCCC4
Molecular Weight (g/mol) 569.65
Formula Weight 569.65
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.