missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 190
GREENER_CHOICE

Product Code. 30002019
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002019 10 mg 10mg
30002023 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002019 Supplier Tocris Bioscience™ Supplier No. 4866/10
  • ONLINE EXCLUSIVE PRICE
    245.00 €
    231.00 € / 10mg
    Save 14.00 €
    5% Off
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective κ opioid receptor antagonist

Selective κ opioid receptor (KOP) antagonist (IC50 = 120 nM in a β-arrestin assay); displays >267-fold selectivity over μ and δ opioid receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1355244-02-8
Molecular Formula C27H32N6O3

Specifications

Quantity 10 mg
Formula Weight 488.58
Molecular Weight (g/mol) 488.592
Chemical Name or Material ML 190
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.