missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 213

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective KV7.2 (KCNQ2) and KV7.4 (KCNQ4) channel opener (EC50 values are 230 and 510 nM for KV7.2 and KV7.4 respectively). Displays > 80-fold selectivity against KV7.1, KV7.3 and KV7.5 in a thallium-based fluorescence assay. Hyperpolarizes resting membrane potential in an in vitro spinal cord preparation.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 257.37 |
| Purity | >99% |
| Molecular Weight (g/mol) | 257.377 |
| Chemical Name or Material | ML 213 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction