CAS RN 489402-47-3
CAS RN 489402-47-3
IUPAC Name:
N-(2,4,6-trimethylphenyl)bicyclo[2.2.1]heptane-2-carboxamide
Synonyms:
N-(2,4,6-trimethylphenyl)bicyclo[2.2.1]heptane-2-carboxamide
Molecular Weight (g/mol):
257.38
Molecular Formula:
C17H23NO
InChi Key:
SIQGKPGBLYKQBB-UHFFFAOYNA-N
SMILES:
CC1=CC(C)=C(NC(=O)C2CC3CCC2C3)C(C)=C1
1
–
2
of
2
results
1
Tocris Bioscience™ ML 213
CAS: 489402-47-3 Molecular Formula: C17H23NO
2
ML213, MedChemExpress
MedChemExpress ML213 is a selective activator of Kv7.2 and Kv7.4 channels, enhances Kv7.2 and Kv7.4 channels with EC50s of 230 and 510 nM, respectively.