missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 213
GREENER_CHOICE

Product Code. 18024232
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18024232

Brand: Tocris Bioscience™ 4519/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KV7.2 and KV7.4 channel opener

Selective KV7.2 (KCNQ2) and KV7.4 (KCNQ4) channel opener (EC50 values are 230 and 510 nM for KV7.2 and KV7.4 respectively). Displays > 80-fold selectivity against KV7.1, KV7.3 and KV7.5 in a thallium-based fluorescence assay. Hyperpolarizes resting membrane potential in an in vitro spinal cord preparation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 489402-47-3
Molecular Formula C17H23NO

Specifications

Quantity 10 mg
Formula Weight 257.37
Purity >99%
Molecular Weight (g/mol) 257.377
Chemical Name or Material ML 213
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.