missing translation for 'onlineSavingsMsg'
Learn More

ML171, MedChemExpress

Product Code. 16481646
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481646 500 mg 500mg
16465986 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481646 Supplier Medchem Express Supplier No. HY12805
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ML171 (2-Acetylphenothiazine;2-APT) is a potent and selective NADPH oxidase 1 (Nox1) inhibitor that blocks Nox1-dependent ROS generation, with an IC50 of 0.25 μM in HEK293-Nox1 confirmatory assay.

ML171 (2-Acetylphenothiazine

Specifications

Chemical Name or Material ML171
CAS 6631-94-3
Color Orange
Purity Grade Notes Research
Molecular Formula C14H11NOS
Quantity 500 mg
Synonym 2-Acetylphenothiazine 2-APT
Solubility Information DMSO : ≥ 64 mg/mL (265.22 mM)
SMILES CC(C1=CC=C2SC3=CC=CC=C3NC2=C1)=O
Molecular Weight (g/mol) 241.31
Formula Weight 241.31
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.