missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ML213, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
ML213 is a selective activator of Kv7.2 and Kv7.4 channels, enhances Kv7.2 and Kv7.4 channels with EC50s of 230 and 510 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | ML213 |
| Physical Form | Solid |
| CAS | 489402-47-3 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H23NO |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 36.67 mg/mL (142.48 mM; Need ultrasonic) |
| SMILES | O=C(C1C(C2)CCC2C1)NC3=C(C)C=C(C)C=C3C |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction