missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLC1SA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99163-50ul
This item is not returnable.
View return policy
Description
MLC1SA Polyclonal antibody specifically detects MLC1SA in Human samples. It is validated for Western Blot
Specifications
| MLC1SA | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MLC1sa, MLC1SAmyosin light chain 1, slow-twitch muscle A isoform, myosin alkali light chain 1 slow a, myosin light chain 1 slow a, Myosin light chain 1 slow-twitch muscle A isoform, myosin light chain 6B, myosin, light chain 6B, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle, smooth muscle and non-muscle myosin alkali light chain 6B, Smooth muscle and nonmuscle myosin light chain alkali 6B | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MLC1SA | |
| 50 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 140465 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction