Learn More
Invitrogen™ MLKL Recombinant Rabbit Monoclonal Antibody (23GB2895), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553520
 View more versions of this product
Product Code. 30088067
  • 562.00 € / 100µL
Description
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MLKL
Recombinant Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NB16, Q9D2Y4
Mlkl
A synthesized peptide derived from human MLKL (350-471AA).
100 μL
Primary
Human, Mouse
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2895
Unconjugated
Mlkl
9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like
Rabbit
Affinity chromatography
RUO
197259, 74568
-20°C