missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MLLT6 Recombinant Rabbit Monoclonal Antibody (K01_2N33)

Product Code. 30827804
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827804 100 μL
30830381 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827804 Supplier Invitrogen™ Supplier No. MA564125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MLLT6 Recombinant Monoclonal Antibody for Western Blot

MLLT6 gene ontology annotations related to this gene include negative regulation of histone H3-K79 methylation; negative regulation of urine volume; positive regulation of sodium ion transport; positive regulation of transcription by RNA polymerase II; renal potassium excretion; renal sodium excretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLLT6
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2N33
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene MLLT6
Gene Accession No. P55198
Gene Alias Af17; AI315037; ALL1-fused gene from chromosome 17 protein; MLLT6; MLLT6, PHD finger domain containing; myeloid/lymphoid or mixed lineage-leukemia translocation to 6 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 6; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6; Myeloid/lymphoid or mixed-lineage leukemia, translocated to, 6; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 6; protein AF-17; trithorax homolog
Gene Symbols MLLT6
Host Species Rabbit
Immunogen Recombinant protein of human MLLT6.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4302
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.