missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLLT6 Recombinant Rabbit Monoclonal Antibody (K01_2N33)
Description
MLLT6 Recombinant Monoclonal Antibody for Western Blot
MLLT6 gene ontology annotations related to this gene include negative regulation of histone H3-K79 methylation; negative regulation of urine volume; positive regulation of sodium ion transport; positive regulation of transcription by RNA polymerase II; renal potassium excretion; renal sodium excretion.
Specifications
Specifications
| Antigen | MLLT6 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2N33 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | MLLT6 |
| Gene Accession No. | P55198 |
| Gene Alias | Af17; AI315037; ALL1-fused gene from chromosome 17 protein; MLLT6; MLLT6, PHD finger domain containing; myeloid/lymphoid or mixed lineage-leukemia translocation to 6 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 6; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6; Myeloid/lymphoid or mixed-lineage leukemia, translocated to, 6; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 6; protein AF-17; trithorax homolog |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?