missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MM-102 TFA, MedChemExpress
Description
MM-102 TFA (HMTase Inhibitor IX TFA) is a potent WDR5/MLL interaction inhibitor, achieves IC50 = 2.4 nM with an estimated Ki 200 times more potent than the ARA peptide.
Specifications
Specifications
| Chemical Name or Material | MM-102 TFA |
| CAS | 1883545-52-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C37H50F5N7O6 |
| Quantity | 50 mg |
| Synonym | HMTase Inhibitor IX TFA |
| Solubility Information | DMSO : ≥ 100 mg/mL (127.58 mM) |
| SMILES | O=C(C1(NC([C@@H](NC(C(NC(C(C)C)=O)(CC)CC)=O)CCCNC(N)=N)=O)CCCC1)NC(C2=CC=C(F)C=C2)C3=CC=C(F)C=C3.FC(F)(F)C(O)=O |
| Molecular Weight (g/mol) | 783.83 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?