missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMAD-d8, MedChemExpress
Description
D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent.
Specifications
Specifications
| Chemical Name or Material | MMAD-d8 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C41H58D8N6O6S |
| Technique | Cancer-programmed Cell Death |
| Color | Off-White |
| Synonym | Demethyldolastatin 10-d8 Monomethylauristatin D-d8 Monomethyl Dolastatin 10-d8 |
| Solubility Information | DMSO : ≥ 100 mg/mL (128.35 mM) |
| SMILES | O=C(N[C@H](C1=NC=CS1)CC2=CC=CC=C2)[C@H](C)[C@@H](OC)[C@H]3N(C(C[C@@H](OC)[C@H]([C@@H](C)CC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C)=O)CCC3 |
| Molecular Weight (g/mol) | 779.11 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?