Learn More
MMP-9 Antibody (CL0538) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52948AF405
Product Code. 30292447
  • 737.00 € / 0.10mL
Description
MMP-9 Monoclonal antibody specifically detects MMP-9 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MMP-9
Monoclonal
Alexa Fluor 405
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
This antibody was developed using a recombinant protein derived from P14780, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
4318
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0538
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified