missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Modified Tryptone Soya Broth with Novobiocin (Bottled)

Product Code. 10609954
Change view
Click to view available options
Fill Weight:
225.0g - 229.0g
Format:
Sirop Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format unitSize
10609954 225.0g - 229.0g Sirop Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10609954 Supplier Thermo Scientific™ Supplier No. BO0869S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Selectively enrich and isolate Escherichia coli O157:H7 from food samples with these ready-prepared bottles.

Selectively enrich and isolate Escherichia coli O157:H7 from food samples with Thermo Scientific™ Oxoid™ Modified Tryptone Soya Broth with Novobiocin. The presence of bile salts and Novobiocin in the medium inhibits the growth of non-target Gram positive organisms. Modified Tryptone Soya Broth is recommended by the U.S. Department of Agriculture (USDA) and the U.S. Food and Drug Administration (FDA) for selective enrichment of certain food samples prior to culture to enhance the recovery of E. coli O157: H71, 2.

E. coli is a part of commensal microbial flora of all animals, including humans. Most strains of E. coli are non-pathogenic and serve a useful function within the intestines by suppressing the growth of harmful bacteria. Some strains of E. coli are capable of causing gastroenteritis in humans, among them is E. coli serotype O157: H7, a rare variety which produces toxins that are closely related to the toxin produced by Shigella dysenteriae1. This toxin causes a severe gastroenteritis in humans commonly referred to as hemorrhagic colitis.

Use Modified Tryptone Soya Broth with Novobiocin for selective enrichment and isolation of E. coli O157:H7 from food samples.

  • Convenient: pre-filled bottles are ready for immediate use removing the need for pipetting, dispensing and autoclaving.
  • Simple: easy-to-use with minimum training.
  • Long shelf life : up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, Clear
Content And Storage 2 to 25°C
Description Modified Tryptone Soya Broth with Novobiocin (Bottled)
Format Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 225.0g - 229.0g
Packaging Type 10 sirop bottles with screw cap closures in a box
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.