missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOG Antibody (CL2858) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46634CL1
This item is not returnable.
View return policy
Description
MOG Monoclonal antibody specifically detects MOG in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen)
Specifications
| MOG | |
| Monoclonal | |
| CoraFluor 1 | |
| MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
| This antibody was developed using a recombinant protein derived from Q16653, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cellular Markers | |
| 4340 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| CL2858 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction