Learn More
MOG Antibody (CL2858) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46634DL594
Product Code. 30303328
  • 791.00 € / 0.10mL
Description
MOG Monoclonal antibody specifically detects MOG in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MOG
Monoclonal
DyLight 594
MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein
This antibody was developed using a recombinant protein derived from Q16653, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers
4340
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
CL2858
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified