missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red Sphingomyelinase Assay Kit

Product Code. 10635013
Change view
Click to view available options
Quantity:
500 Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10635013 500 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10635013 Supplier Invitrogen™ Supplier No. A12220
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detects very low concentrations of sphingomyelinase

The Amplex™ Red Sphingomyelinase Assay Kit provides a sensitive, rapid, and simple fluorometric method for detecting very low concentrations of sphingomyelinase using a fluorescence microplate reader or fluorometer.

  • Detects sphingomyelinase activity levels as low as 80 μU/mL in a 200 μL assay volume
  • Format allows for multiple time point measurements
  • Designed for minimal autofluorescence interference

In this enzyme-coupled assay, sphingomyelinase activity is monitored indirectly using 10-acetyl-3,7-dihydroxyphenoxazine (Amplex™ Red reagent), a sensitive fluorogenic probe for hydrogen peroxide. Sphingomyelinase hydrolyses the sphingomyelin to yield ceramide and phosphorylcholine. Alkaline phosphatase is added, which hydrolyzes phosphorylcholine to form choline. The choline is then oxidized by choline oxidase to form betaine and hydrogen peroxide. In the presence of horseradish peroxidase, the hydrogen peroxide reacts with Amplex™ Red reagent in a 1:1 stoichiometric ratio to generate the highly fluorescent product resorufin.

Because resorufin has absorption and fluorescence emission maxima of approximately 571 nm and 585 nm, respectively, there is little interference from autofluorescence in most biological samples.

Use Amplex™ Red Assays for a Broad Range of Investigations

A wide variety of validated Amplex™ Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex™ UltraRed Reagent (Cat. No. A36006), a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex™ Red/UltraRed Stop Reagent (Cat. No. A33855). The Amplex™ Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours. Custom assay design and packaging are also available.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Enzyme and Metabolic Activity
For Use With (Equipment) Fluorometer, Microplate Reader
Product Type Amplex Red Assay Kit
Dye Type Amplex Red
Excitation Wavelength Range 570 to 585 nm
Format 96-well Plate
Product Line Amplex
Quantity 500 Assays
Shipping Condition Wet Ice
Emission Wavelength Range 570 to 585 nm, 585 to 600 nm
Excitation Wavelength 571 nm
Emission Wavelength 585 nm
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Amplex Red Sphingomyelinase Assay Kit
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, .alpha.-[4-(1,1,3,3-tetramethylbutyl)phenyl]-.omega.-hydroxypoly(oxy-1,2-ethanediyl)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.