missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAPTA, AM, cell permeant chelator

A cell-permeant chelator, which is a highly selective for Ca2+ over Mg2+

Brand:  Invitrogen™ B6769

Additional Details : Weight : 0.00525kg

 View more versions of this product

Product Code. 10022612

  • 475.00 € / 20mg
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Can be used to control the level of intracellular Ca2+
  • BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Specifications

Specifications

Store in freezer -5°C to -30°C.
Cell Viability & Proliferation
20 x 1mg
Cell Viability & Proliferation
Chelator (Non-Fluorescent)
Room Temperature
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.