missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calcium Green™-1 dextran, Potassium Salt, 10,000 MW, Anionic

Product Code. 11528946
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11528946 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11528946 Supplier Invitrogen™ Supplier No. C3713
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues. Cells may be physically loaded with the cell-impermeant salt forms of these dextran-conjugated indicators using patch pipette, microinjection, or our Influx pinocyte-loading reagent (Cat. No. I14402). The fluorescence signal from these cells is measured using fluorescence microscopy. The dextran forms of our calcium indicators show a dramatic reduction in both leakage and compartmentalization compared to the AM ester forms.

Calcium Indicator (Cell-Impermeant Salts, Dextran Conjugates) Specifications:

  • Label (Ex/Em): Calcium Green™ -1 (506/531 nm)
  • Fluorescence intensity increase upon binding Ca2+: ∼14 fold
  • Dextran size: 10,000 MW
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

Spectral Characteristics of Molecular Probes™ Calcium Indicators

These probes are excited by visible light, and because the energy required for excitation is low, the potential for cellular photodamage is reduced. Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of the spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem.

More Choices for Fluorescent Calcium Indicators

We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios, for example dextran versions for reduced leakage and compartmentalization and BAPTA conjugates for detecting high-amplitude calcium transients. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Calcium Indicator
For Use With (Equipment) Fluorescence Microscope
Product Type Calcium Indicator
Dye Type Calcium Green
Excitation Wavelength Range 495 to 510 nm
Product Line Calcium Green
Quantity 5 mg
Shipping Condition Room Temperature
Emission Wavelength Range 525 to 540 nm
Excitation Wavelength 507 nm
Emission Wavelength 531 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.