missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Gelatinase/Collagenase Assay Kit, 250-2,000 assays

Product Code. 10104812
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10104812 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10104812 Supplier Invitrogen™ Supplier No. E12055
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Fluorometric method for measuring gelatinase or collagenase activity

The EnzChek Gelatinase/Collagenase Assay Kit provides a sensitive, convenient and fast fluorometric method for measuring gelatinase or collagenase activity in purified enzyme systems, cell/tissue lysates or for screening inhibitors in a high-throughput format. The substrate in the EnzChek kit is fluorescein-labeled DQ gelatin conjugate that is highly labeled so that the fluorescence signal is quenched until enzymatic digestion yields highly fluorescent fragments. This substrate is also available separate from the kit (D-12054).

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Chemotaxis and Cell Migration, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Gelatinase/Collagenase Assay
Product Type EnzChek Assay Kit
Product Line EnzChek
Quantity 1 Kit
Shipping Condition Room Temperature
Substrate Properties Protein-Based Substrate
Substrate Type Gelatinase/Collagenase Substrate
Target Enzyme Gelatinase/Collagenase

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.