missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Lysozyme Assay Kit

Product Code. 10565393
Change view
Click to view available options
Quantity:
400 Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10565393 400 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10565393 Supplier Invitrogen™ Supplier No. E22013
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Measure lysozyme activity in solution

The EnzChek Lysozyme Assay Kit provides researchers with a simple and sensitive assay to measure lysozyme activity in solution. The increase in fluorescence resulting from lysozyme activity is measured using a fluorometer or fluorescence microplate reader. The assay utilizes an innovative approach to measure lysozyme activity. Micrococcus lysodeikticus cell walls are used that have been extensively labeled with fluorescein causing quench of the fluorescence signal. Active lysozyme enzyme hydrolyzes the b-(1-4)-glucosidic linkages between the N-acetylmuramic acid and N-acetyl-D-glucosamine residues in the mucopolysaccharide cell wall, relieving the quenching and yielding a dramatic increase in fluorescence that is proportional to lysozyme activity.

  • Detect lysozyme activity concentrations as low as 20U/mL
  • Use standard fluorescein (FITC) excitation/emission settings
  • Format allows for continuous measurement of lysozyme activity

Cell Analysis, Cellular Imaging, Enzyme and Protein Activity Assays, Glycosidase, Neuraminidase, and Related Enzyme Activity, High-Content Screening (HCS), Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Lysozyme Assay
For Use With (Equipment) Fluorescence Microplate Reader
Product Type EnzChek Assay Kit
Product Line EnzChek
Quantity 400 Assays
Shipping Condition Room Temperature
Substrate Properties Lipid-Based Substrate
Substrate Type Lysozyme Substrate
Target Enzyme Lysozyme
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.