missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoZin™-3, AM, cell permeant

Product Code. 11504816
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 11504816

Brand: Invitrogen™ F24195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zn2+-selective indicator with a structure that resembles fluo-4

  • Exhibits high Zn2+-binding affinity that is unperturbed by Ca2+ concentrations up to at least 1μM
  • Exhibits >50-fold increase in fluorescence in response to saturating levels of Zn2+
  • Although imaging applications of FluoZin-3 are predominant, cell-based microplate assays and flow cytometry protocols have also been developed
  • Label (Ex/Em): FluoZin-3 (approximately 494/516nm)
  • Kd (Zn2+) (in buffer): approximately 15 nM
  • Lyophilized product may be dissolved in DMSO for use
  • Product is typically loaded into cells by adding the dissolved indicator to medium containing cells

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Drug Discovery and Development, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation, Zinc and Heavy Metal Ion Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type Zinc Indicator
Dye Type Zinc Indicator
Product Line FluoZin
Quantity 100 μg
Shipping Condition Room Temperature
Molecular Weight (g/mol) 982.85 g/mol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.