missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fura-4F, AM, cell permeant - Special Packaging

Product Code. 11534776
Change view
Click to view available options
Quantity:
10 x 50 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11534776 10 x 50 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11534776 Supplier Invitrogen™ Supplier No. F14175

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Intracellular calcium indicator similar to fura-2 but with a response range shifted to detect higher Ca2+ concentrations

  • Acetoxymethyl (AM) ester derivative
  • Useful for noninvasive intracellular loading
  • Ex/Ems are identical to that of fura-2, and more importantly the Kd (Ca2+)μM for each are very similar: 0.77 for fura-4F AM

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Cell-Based Second Messenger Assays, Drug Discovery and Development, G-Protein Coupled Receptor Biology, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type Dye
Dye Type Fluorescent Dye-Based
Quantity 10 x 50 μg
Shipping Condition Room Temperature
I am doing calcium flux imaging with your Fura-2 calibration kit, but am seeing a large variability in ratio in different places around the slide. I am correcting for uniform illumination, using the product as directed, and sealing the coverslip with nail polish.

The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).

I need to label cells with Fluo-4, AM, for a calcium flux assay. How long after labeling will the dye be retained?

After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.