missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mag-Fura-2, Tetrapotassium Salt, cell impermeant

Product Code. 10403272
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10403272 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10403272 Supplier Invitrogen™ Supplier No. M1290
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Intracellular magnesium indicator that is ratiometric and UV light-excitable.

Mag-fura-2, tetrapotassium salt is an intracellular magnesium indicator that is ratiometric and UV light-excitable. This water-soluble salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (I14402).

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Magnesium Indicator
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Magnesium Indicator
Dye Type Mag-Fura-2
Excitation Wavelength Range 330 to 345 nm
Quantity 1 mg
Shipping Condition Room Temperature
Emission Wavelength Range 390 to 405 nm
Excitation Wavelength 330 nm
Emission Wavelength 491 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.