missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PIP Strips™ Membranes

Product Code. 10768044
Click to view available options
Quantity:
10 strips
2 strips
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10768044

Brand: Invitrogen™ P23750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identify proteins possessing phosphoinositide domains

PIP Strips membranes are designed for the identification of proteins possessing phosphoinositide domains and for analysis of their lipid-binding specificities. PIP Strips membranes facilitate the analysis of phosphoinositide protein interactions by protein-lipid overlay assays. Proteins may be detected using standard western blot procedures in conjunction with high-performance alkaline phosphatase- and horseradish peroxidase (HRP)-mediated signal generation systems.

Cell Analysis, Cell Structure, Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Membranes (General) and Lipids, Plasma Membrane, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Product Type Phospholipid Arrays
Format Membrane
Product Line PIP Strips
Quantity 2 strips
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.