missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rhod-5N, Tripotassium Salt, cell impermeant

Product Code. 11563677
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11563677 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11563677 Supplier Invitrogen™ Supplier No. R14207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cells may be physically loaded with the cell-impermeant salt forms of these indicators using patch pipet, microinjection or pinocytotic cell-loading reagent

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring Ca2+ in cells and tissues that have high levels of autofluorescence and also for detecting Ca2+ release generated by photoreceptors and photoactivatable chelators.

  • Fluorescence signal from these cells is generally measured using fluorescence microscopy
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in absence of Mg2+, in buffer: approximately 320μM
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
  • In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+
  • Perturbations to calcium measurements caused by presence of these ions can be controlled using heavy metal-selective chelator TPEN

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Quantity 500 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.