Learn More
Invitrogen™ Rhodamine 123, 25 mg
Description
Rhodamine 123 is readily sequestered by active mitochondria without cytotoxic effects. This product has been used to assay mitochondrial membrane potential in populations of apoptotic cells.
Apoptosis, Cell Analysis, Cell Structure, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Mitochondria, Mitochondria Function, Mitochondrial Function, Mitochondrial Structure
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Green |
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Rhodamine 123 |
| Quantity | 25 mg |
| Shipping Condition | Room Temperature |
| Sub Cellular Localization | Mitochondria |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.