missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tetramethylrhodamine, Ethyl Ester, Perchlorate (TMRE)

Product Code. 11560796
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11560796 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11560796 Supplier Invitrogen™ Supplier No. T669
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tetramethylrhodamine, ethyl ester (TMRE) is a cell-permeant, cationic, red-orange fluorescent dye that is readily sequestered by active mitochondria.

Tetramethylrhodamine, ethyl ester (TMRE) is a cell-permeant, cationic, red-orange fluorescent dye that is readily sequestered by active mitochondria.

Dissolve the dye in high-quality anhydrous DMSO or ethanol to prepare stock concentration at 1 to 10 mM.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red-Orange
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type TMRE
Quantity 25 mg
Shipping Condition Room Temperature
Sub Cellular Localization Mitochondria
Excitation/Emission 549/574 nm
Molecular Weight (g/mol) 514.96

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.