missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vybrant™ Phagocytosis Assay Kit
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kit
Unit Size:
Each
Description
- Foreign particle is killed, E. coli (K-12 strain) cells that have been labeled with fluorescent dye fluorescein
- Addition to lyophilized E. coli BioParticles™ component, kit contains a trypan blue solution (to quench fluorescence from particles that were not internalized) as well as step-by-step instructions for performing this phagocytosis assay in a fluorescence microplate reader
- Methodology used with this kit has been developed using an adherent murine macrophage cell line (J774); however, by modifying the cell culture conditions, researchers can adapt this phagocytosis assay to other adherent cell lines
- Particles are fluorescein-labeled E. coli (K-12 strain)
- Kit supplies sufficient reagents for approx. 250 tests using 96-well plates
- Fluorescence is measured using a microplate reader (Ex/Em approx. 480/520nm)
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Microplate Reader |
| Product Type | Phagocytosis Assay Kit |
| Dye Type | Classic Dyes |
| Format | 96-well plate |
| Product Line | Vybrant |
| Quantity | 1 kit |
| Shipping Condition | Room Temperature |
| Species | E. coli |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction